Alfa Aesar™ Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV), 99.7% (metals basis)

Manufacturer: Alfa Aesar™ 4190903

Quantity 1g

 View more versions of this product

Catalog No. AA4190903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 63370-90-1
    Formula Weight 910.5
    MDL Number MFCD01863471
    Synonym Hafnium tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionate); Hf(TMHD)4
    Chemical Name or Material Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV)

    Specifications

    Formula Weight 910.5
    Melting Point 315°C
    Physical Form Powder
    Synonym Hafnium tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionate); Hf(TMHD)4
    MDL Number MFCD01863471
    CAS 63370-90-1
    Quantity 1g
    Chemical Name or Material Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV)
    Assay 99.7%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit