Alfa Aesar™ Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)niobium(IV)

Manufacturer: Alfa Aesar™ 3945906

Quantity 5g

 View more versions of this product

Catalog No. AA3945906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 41706-15-4
    Formula Weight 826
    MDL Number MFCD00145379
    Synonym Niobium tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionate); Nb(TMHD)4
    Chemical Name or Material Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)niobium(IV)

    Specifications

    Formula Weight 826
    Physical Form Powder
    CAS 41706-15-4
    Synonym Niobium tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionate); Nb(TMHD)4
    Melting Point 219° to 220°C
    Quantity 5g
    MDL Number MFCD00145379
    Boiling Point 325°C (dec.)
    Chemical Name or Material Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)niobium(IV)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit