Alfa Aesar™ Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), 99.99% (metals basis)

MOCVD of zirconium

Manufacturer: Alfa Aesar™ 4202803

Quantity 1g

 View more versions of this product

Catalog No. AA4202803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18865-74-2
    Formula Weight 824.3
    MDL Number MFCD00145380
    Synonym Zr(TMHD)4
    Chemical Name or Material Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV)

    Specifications

    Formula Weight 824.3
    Synonym Zr(TMHD)4
    Boiling Point 180°C (0.1mmHg) (subl.)
    Melting Point 318° to 320°C
    Quantity 1g
    Physical Form Crystalline
    MDL Number MFCD00145380
    CAS 18865-74-2
    Chemical Name or Material Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV)
    Assay 99.99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit