Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Tetrakis(acetonitrile)palladium(II) tetrafluoroborate, Pd 23.5%
Click to view available options
Quantity:
0.25g
1g
Specifications
Concentration or Composition (by Analyte or Components) | Pd 23.5% |
Sensitivity | Air Sensitive |
Quantity | 1g |
Melting Point | 230°C (decomposition) |
UN Number | UN2811 |
Solubility Information | Soluble in water |
Formula Weight | 444.22 |
Physical Form | Powder |
Chemical Name or Material | Tetrakis(acetonitrile)palladium(II) tetrafluoroborate |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P271-P304+P340-P312
H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures