Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetrakis(diethylamino)titanium(IV), 99.99% (metals basis)

Nitride precursor - MOCVD

Manufacturer:  Alfa Aesar™ 4046918

Quantity 50g

 View more versions of this product

Catalog No. AA4046918


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

4419-47-0
336.4
UN3129
Tetrakis(diethylamino)titanium(IV)
Ti[N(C2H5)2]4
MFCD00015648
Titanium diethylamide

Specifications

4419-47-0
Liquid
336.4
UN3129
1.5370
Orange to red
Air & Moisture Sensitive
99.99%, (metals basis)
0.95g/mL
Ti[N(C2H5)2]4
MFCD00015648
Titanium diethylamide
133°C (1.2mmHg)
50g
Tetrakis(diethylamino)titanium(IV)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit