Alfa Aesar™ Tetrakis(diethylamino)titanium(IV), 99.99% (metals basis)

Nitride precursor - MOCVD

Manufacturer: Alfa Aesar™ 4046904

Quantity 2g

 View more versions of this product

Catalog No. AA4046904

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4419-47-0
    Linear Formula Ti[N(C2H5)2]4
    Formula Weight 336.4
    MDL Number MFCD00015648
    UN Number UN3129
    Synonym Titanium diethylamide
    Chemical Name or Material Tetrakis(diethylamino)titanium(IV)

    Specifications

    Formula Weight 336.4
    Health Hazard 3 GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    UN Number UN3129
    Health Hazard 1 Danger
    Refractive Index 1.5370
    Health Hazard 2 GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Quantity 2g
    Density 0.95g/mL
    Linear Formula Ti[N(C2H5)2]4
    Boiling Point 133°C (1.2mmHg)
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.
    MDL Number MFCD00015648
    CAS 4419-47-0
    Synonym Titanium diethylamide
    Sensitivity Air & Moisture Sensitive
    TSCA TSCA
    Color Orange to red
    Chemical Name or Material Tetrakis(diethylamino)titanium(IV)
    Assay 99.99%, (metals basis)

    GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit