Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetrakis(dimethylamino)germanium(IV), 99% (metals basis)

Preparation of nitrides

Manufacturer:  Alfa Aesar™ 42760ME

Quantity 500mg

 View more versions of this product

Catalog No. AA42760ME


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7344-40-3
Ge[N(CH3)2]4
MFCD01863476
Germanium dimethylamide
C8H24GeN4
248.9
UN3129
Tetrakis (dimethylamino)germanium (IV)

Specifications

203°C
7344-40-3
34°C (93°F)
Colorless
Moisture Sensitive
C8H24GeN4
248.9
UN3129
Tetrakis (dimethylamino)germanium (IV)
14°C
500mg
1.07g/mL
Amine-like
Liquid
Ge[N(CH3)2]4
MFCD01863476
Germanium dimethylamide
99%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H261-H226-H314-H318
In contact with water releases flammable gas.
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit