Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(dimethylamino)titanium, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3275-24-9
C8H24N4Ti
224.21
MFCD00014861
MNWRORMXBIWXCI-UHFFFAOYSA-N
tetrakis dimethylamino titanium,tetrakis dimethylamino titanium iv,tdmat,tetrakis dimethylamido titanium iv,tetrakis dimethylamido titanium,methanamine, n-methyl-, titanium 4+ salt,titanium dimethylamide,methanamine, n-methyl-, titanium 4+ salt 4:1,titanium tetrakis dimethylamide,acmc-209hva
123185
dimethylazanide;titanium(4+)
C[N-]C.C[N-]C.C[N-]C.C[N-]C.[Ti+4]
3275-24-9
C8H24N4Ti
224.21
MFCD00014861
MNWRORMXBIWXCI-UHFFFAOYSA-N
tetrakis dimethylamino titanium,tetrakis dimethylamino titanium iv,tdmat,tetrakis dimethylamido titanium iv,tetrakis dimethylamido titanium,methanamine, n-methyl-, titanium 4+ salt,titanium dimethylamide,methanamine, n-methyl-, titanium 4+ salt 4:1,titanium tetrakis dimethylamide,acmc-209hva
123185
dimethylazanide;titanium(4+)
C[N-]C.C[N-]C.C[N-]C.C[N-]C.[Ti+4]
For ultra trace analysis
Flash Point -30°C
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 0.947
Quantity 5g
Density 0.9470g/mL
Color Yellow to Orange
Assay Percent Range 99.99%
Linear Formula [(CH3)2N]4Ti
Beilstein 04, IV, 128
Formula Weight 224.21
Physical Form Liquid
Percent Purity 99.99%
Grade Trace Metal Basis
Chemical Name or Material Tetrakis(dimethylamino)titanium
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger