Alfa Aesar™ Tetrakis(dimethylamino)titanium(IV), 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4293203

Quantity 1g

 View more versions of this product

Catalog No. AA4293203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3275-24-9
    Molecular Formula C8H24N4Ti
    Linear Formula Ti[N(CH3)2]4
    Formula Weight 224.19
    MDL Number MFCD00014861
    UN Number UN3129
    Synonym Titanium dimethylamide
    Chemical Name or Material Tetrakis (dimethylamino)titanium (IV)

    Specifications

    Formula Weight 224.19
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Density 0.96g/mL
    Linear Formula Ti[N(CH3)2]4
    MDL Number MFCD00014861
    Quantity 1g
    Molecular Formula C8H24N4Ti
    CAS 3275-24-9
    UN Number UN3129
    Flash Point −30°C (−22°F)
    Sensitivity Moisture Sensitive
    Synonym Titanium dimethylamide
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H261-H314-H318
    Highly flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Boiling Point 50°C (0.5mmHg)
    Color Yellow-brown
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.
    Physical Form Liquid
    Chemical Name or Material Tetrakis (dimethylamino)titanium (IV)
    Assay 99.9%, (metals basis)

    GHS H Statement
    H225-H261-H314-H318
    Highly flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit