Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(dimethylamino)zirconium, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
19756-04-8
C8H24N4Zr
267.53
MFCD00239502
DWCMDRNGBIZOQL-UHFFFAOYSA-N
tetrakis dimethylamino zirconium,zirconium, tetrakis dimethylamino,tdmaz,tetrakis dimethylamino zirconium iv,tetrakis dimethylamido zirconium iv,zr nme2 4,dimethyl tris dimethylamino zirconio amine,tetrakis dimethylamido zirconium iv , crystalline,tetrakis dimethylamido zirconium iv , deposition grade,tetrakis dimethylamido zirconium iv , packaged for use in deposition systems
140580
dimethylazanide;zirconium(4+)
C[N-]C.C[N-]C.C[N-]C.C[N-]C.[Zr+4]
19756-04-8
C8H24N4Zr
267.53
MFCD00239502
DWCMDRNGBIZOQL-UHFFFAOYSA-N
tetrakis dimethylamino zirconium,zirconium, tetrakis dimethylamino,tdmaz,tetrakis dimethylamino zirconium iv,tetrakis dimethylamido zirconium iv,zr nme2 4,dimethyl tris dimethylamino zirconio amine,tetrakis dimethylamido zirconium iv , crystalline,tetrakis dimethylamido zirconium iv , deposition grade,tetrakis dimethylamido zirconium iv , packaged for use in deposition systems
140580
dimethylazanide;zirconium(4+)
C[N-]C.C[N-]C.C[N-]C.C[N-]C.[Zr+4]
Melting Point 57°C to 60°C
Color Green-Yellow
Packaging Glass bottle
Quantity 1g
NMR Structure confirmed
Formula Weight 267.53
Physical Form Crystalline Solid
Chemical Name or Material Tetrakis(dimethylamino)zirconium

GHS H Statement
Flammable solid.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye prot

GHS Signal Word: Danger