Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrakis(dimethylsiloxy)silane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It is employed for non-volatile NMR standard for high temperature work, in place of the highly volatile and as cross linker for vinyl functional silicones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed for non-volatile NMR standard for high temperature work, in place of the highly volatile and as cross linker for vinyl functional silicones.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
It is employed for non-volatile NMR standard for high temperature work, in place of the highly volatile and as cross linker for vinyl functional silicones.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Specifications
Boiling Point | 188°C to 190°C |
Melting Point | -60°C |
Quantity | 100 g |
Beilstein | 1793898 |
Flash Point | 67°C (153°F) |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 328.73 |
Percent Purity | 97% |
Refractive Index | 1.388 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H227-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction