Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(hydroxymethyl)phosphonium Chloride (ca. 80% in Water), TCI America™

Click to view available options
Quantity:
25mL
500mL
124-64-1
C4H12ClO4P
190.56
MFCD00031687
AKXUUJCMWZFYMV-UHFFFAOYSA-M
tetrakis hydroxymethyl phosphonium chloride,thpc,pyroset tkc,retardol c,tetramethylolphosphonium chloride,tetrakis hydroxymethyl phosphanium chloride,unii-58wb2xcf8i,proban cc,tetrakis hydroxymethyl phosphochloride,ccris 317
31298
tetrakis(hydroxymethyl)phosphanium;chloride
C(O)[P+](CO)(CO)CO.[Cl-]
124-64-1
C4H12ClO4P
190.56
MFCD00031687
AKXUUJCMWZFYMV-UHFFFAOYSA-M
tetrakis hydroxymethyl phosphonium chloride,thpc,pyroset tkc,retardol c,tetramethylolphosphonium chloride,tetrakis hydroxymethyl phosphanium chloride,unii-58wb2xcf8i,proban cc,tetrakis hydroxymethyl phosphochloride,ccris 317
31298
tetrakis(hydroxymethyl)phosphanium;chloride
C(O)[P+](CO)(CO)CO.[Cl-]
Quantity 500mL
Color Colorless
pH 3.5 - 4.5
UN Number 2922
Formula Weight 190.56
Physical Form Clear Liquid at 20°C
Chemical Name or Material Tetrakis(hydroxymethyl)phosphonium Chloride (ca. 80% in Water)

EINECSNumber : (2)-2003

RTECSNumber : TA2450000

TSCA : Yes