Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(trimethylsilyloxy)silane, 96%, Acros Organics™

Click to view available options
Quantity:
5g
3555-47-3
C12H36O4Si5
384.84
MFCD00051587
VNRWTCZXQWOWIG-UHFFFAOYSA-N
tetrakis trimethylsiloxy silane,tetrakis trimethylsilyl orthosilicate,tetrakis trimethylsilyloxy silane,unii-55l9t9a11i,tetrakis trimethylsilyl silicate,trisiloxane, 1,1,1,5,5,5-hexamethyl-3,3-bis trimethylsiloxy,trisiloxane, 1,1,1,5,5,5-hexamethyl-3,3-bis trimethylsilyl oxy,acmc-1cnqp,tetratrimethylsilyl silicate
19086
tetrakis(trimethylsilyl) silicate
C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
3555-47-3
C12H36O4Si5
384.84
MFCD00051587
VNRWTCZXQWOWIG-UHFFFAOYSA-N
tetrakis trimethylsiloxy silane,tetrakis trimethylsilyl orthosilicate,tetrakis trimethylsilyloxy silane,unii-55l9t9a11i,tetrakis trimethylsilyl silicate,trisiloxane, 1,1,1,5,5,5-hexamethyl-3,3-bis trimethylsiloxy,trisiloxane, 1,1,1,5,5,5-hexamethyl-3,3-bis trimethylsilyl oxy,acmc-1cnqp,tetratrimethylsilyl silicate
19086
tetrakis(trimethylsilyl) silicate
C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
Specific Gravity 0.868
Flash Point 80°C
Density 0.8680g/mL
Boiling Point 103.0°C to 106.0°C (2.0 mmHg)
Packaging Glass bottle
Quantity 5g
Melting Point -60.0°C
Assay Percent Range 95% min. (GC)
Linear Formula Si[OSi(CH3)3]4
Formula Weight 384.84
Percent Purity 96%
Chemical Name or Material Tetrakis(trimethylsilyloxy)silane, 98%
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 222-613-4

TSCA : TSCA