Alfa Aesar™ Tetrakis(triphenylphosphine)nickel(0)

Manufacturer: Alfa Aesar™ 3210103

Quantity 1g

 View more versions of this product

Catalog No. AA3210103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15133-82-1
    Linear Formula Ni[P(C6H5)3]4
    Formula Weight 1107.83
    MDL Number MFCD00010011
    UN Number UN1325
    Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)

    Specifications

    Formula Weight 1107.83
    Health Hazard 3 GHS P Statement
    P210-P201-P309-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Linear Formula Ni[P(C6H5)3]4
    MDL Number MFCD00010011
    CAS 15133-82-1
    UN Number UN1325
    Sensitivity Air & Moisture Sensitive
    Physical Form Crystalline
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H350-H372-H228-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Flammable solid.
    May cause an allergic skin reaction.
    Color Brown
    Melting Point 123° to 128°C
    Quantity 1g
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)

    GHS H Statement
    H350-H372-H228-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Flammable solid.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P201-P309-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit