Alfa Aesar™ Tetrakis(triphenylphosphine)nickel(0)

Manufacturer: Alfa Aesar™ 32101MD

Quantity 250mg

 View more versions of this product

Catalog No. AA32101MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15133-82-1
    Linear Formula Ni[P(C6H5)3]4
    Formula Weight 1107.83
    MDL Number MFCD00010011
    UN Number UN1325
    Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)

    Specifications

    Formula Weight 1107.83
    Health Hazard 3 GHS P Statement
    P210-P201-P309-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Linear Formula Ni[P(C6H5)3]4
    MDL Number MFCD00010011
    CAS 15133-82-1
    Physical Form Crystalline
    UN Number UN1325
    Health Hazard 1 Danger
    Color Brown
    Melting Point 123° to 128°C
    Sensitivity Air & Moisture Sensitive
    Health Hazard 2 GHS H Statement
    H350-H372-H228-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Flammable solid.
    May cause an allergic skin reaction.
    Quantity 250mg
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)

    GHS H Statement
    H350-H372-H228-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    Flammable solid.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P201-P309-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit