Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrakis(triphenylphosphine)palladium(0), 99,8% (metals basis), Pd 9% min, Alfa Aesar™
Click to view available options
Quantity:
0.5g
2g
10g
Specifications
Quantity | 0.5g |
Sensitivity | Air sensitive |
Melting Point | 100°C to 105°C |
Assay Percent Range | 99.8% (metals basis), Pd 9% min |
Beilstein | 6704828 |
Solubility Information | Insoluble in water. Soluble in benzene,ethanol and chloroform |
Formula Weight | 1155.57 |
Physical Form | Crystalline |
Chemical Name or Material | Tetrakis(triphenylphosphine)palladium |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302-H500
EINECSNumber : 238-086-9
TSCA : Yes
storageNote1 : Air Sensitive
Recommended Storage : Store at -20°C; Store under Argon