Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(triphenylphosphine)palladium(0), 99.9%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
1g
5g
14221-01-3
C72H60P4Pd
1155.58
NFHFRUOZVGFOOS-UHFFFAOYSA-N
tetrakis triphenylphosphine palladium,tetrakis triphenylphosphine palladium 0,pd pph3 4,tetrakis triphenylphosphine palladium o,tetra triphenylphosphine palladium,palladium-tetrakis triphenylphosphine,palladium 0 tetrakis triphenylphosphine,palladium, tetrakis triphenylphosphine-, t-4
11979704
palladium;triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pd]
14221-01-3
C72H60P4Pd
1155.58
NFHFRUOZVGFOOS-UHFFFAOYSA-N
tetrakis triphenylphosphine palladium,tetrakis triphenylphosphine palladium 0,pd pph3 4,tetrakis triphenylphosphine palladium o,tetra triphenylphosphine palladium,palladium-tetrakis triphenylphosphine,palladium 0 tetrakis triphenylphosphine,palladium, tetrakis triphenylphosphine-, t-4
11979704
palladium;triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pd]
For ultra trace analysis
Total Trace Metal Impurities 0.1% max
Melting Point 103.0°C to 107.0°C
Quantity 1g
Packaging Glass bottle
Assay Percent Range 99.9%
Linear Formula [(C6H5)3P]4Pd
Merck Index 15,9367
Solubility Information Solubility in water: insoluble.
Formula Weight 1155.58
Percent Purity 99.9%
Grade Trace Metal Basis
Chemical Name or Material Tetrakis(triphenylphosphine)palladium(0)
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 238-086-9