Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(triphenylphosphine)palladium(0), 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
100g
Packaging:
Glass bottle
14221-01-3
C72H60P4Pd
1155.58
NFHFRUOZVGFOOS-UHFFFAOYSA-N
tetrakis triphenylphosphine palladium,tetrakis triphenylphosphine palladium 0,pd pph3 4,tetrakis triphenylphosphine palladium o,tetra triphenylphosphine palladium,palladium-tetrakis triphenylphosphine,palladium 0 tetrakis triphenylphosphine,palladium, tetrakis triphenylphosphine-, t-4
11979704
palladium;triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pd]
14221-01-3
C72H60P4Pd
1155.58
NFHFRUOZVGFOOS-UHFFFAOYSA-N
tetrakis triphenylphosphine palladium,tetrakis triphenylphosphine palladium 0,pd pph3 4,tetrakis triphenylphosphine palladium o,tetra triphenylphosphine palladium,palladium-tetrakis triphenylphosphine,palladium 0 tetrakis triphenylphosphine,palladium, tetrakis triphenylphosphine-, t-4
11979704
palladium;triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Pd]
Packaging Glass bottle
Color Khaki to Yellow
Melting Point 103.0°C to 107.0°C
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 99%
Linear Formula [(C6H5)3P]4Pd
Fieser 06,571; 07,357; 08,472; 09,451; 10,384; 11,503; 12,468; 13,34; 14,177; 16,98; 17,327
Merck Index 15,9367
Solubility Information Solubility in water: insoluble.
Formula Weight 1155.58
Physical Form Fine Powder or Platelets
Percent Purity 99%
Chemical Name or Material Tetrakis(triphenylphosphine)palladium(0)
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 238-086-9