Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium Borohydride 90.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
16883-45-7
C4H12BN
84.957
MFCD00011778
TVQBZLPZXCVNEO-UHFFFAOYSA-N
tetramethylammonium borohydride,borohydride, polymer-supported,tetramethylammonium borohydrate,tetramethylammonium tetrahydroborate 1-,tetramethylammonium tetrahydroborate,tetra-methyl ammonium boron hydride,boron 1-; tetramethylazanium,n,n,n-trimethylmethanaminium tetrahydroborate 1-,ammonium, tetramethyl-, tetrahydroborate 1-,methanaminium, n,n,n-trimethyl-, tetrahydroborate 1-
6328186
boron(1-);tetramethylazanium
[B-].C[N+](C)(C)C
16883-45-7
C4H12BN
84.957
MFCD00011778
TVQBZLPZXCVNEO-UHFFFAOYSA-N
tetramethylammonium borohydride,borohydride, polymer-supported,tetramethylammonium borohydrate,tetramethylammonium tetrahydroborate 1-,tetramethylammonium tetrahydroborate,tetra-methyl ammonium boron hydride,boron 1-; tetramethylazanium,n,n,n-trimethylmethanaminium tetrahydroborate 1-,ammonium, tetramethyl-, tetrahydroborate 1-,methanaminium, n,n,n-trimethyl-, tetrahydroborate 1-
6328186
boron(1-);tetramethylazanium
[B-].C[N+](C)(C)C
Quantity 25g
Color White
UN Number 3131
Formula Weight 88.99
Physical Form Crystal-Powder at 20°C
Percent Purity ≥90.0% (T)
Chemical Name or Material Tetramethylammonium Borohydride [Reducing Reagent]

EINECSNumber : (2)-0186

RTECSNumber : BS8310000

TSCA : Yes