Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetramethylammonium hexafluorophosphate, 99+%

Click to view available options
Quantity:
25g
100g
500g
558-32-7
C4H12F6NP
219.111
MFCD00012012
ZFKULDQWLXAKIM-UHFFFAOYSA-N
tetramethylammonium hexafluorophosphate,ammonium, tetramethyl-, hexafluorophosphate 1-,tetramethylazanium hexafluorophosphate,c4h12n.f6p,acmc-209lpp,hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion,methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-,methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci
11209
tetramethylazanium;hexafluorophosphate
C[N+](C)(C)C.F[P-](F)(F)(F)(F)F
558-32-7
C4H12F6NP
219.111
MFCD00012012
ZFKULDQWLXAKIM-UHFFFAOYSA-N
tetramethylammonium hexafluorophosphate,ammonium, tetramethyl-, hexafluorophosphate 1-,tetramethylazanium hexafluorophosphate,c4h12n.f6p,acmc-209lpp,hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion,methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-,methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci
11209
tetramethylazanium;hexafluorophosphate
C[N+](C)(C)C.F[P-](F)(F)(F)(F)F
Applications
It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
Sensitivity Moisture sensitive
Odor Odorless
Quantity 500g
Melting Point >300°C
Linear Formula (CH3)4NPF6
UN Number UN2923
Beilstein 3730394
Solubility Information Soluble in water.
Formula Weight 219.1
Percent Purity ≥99%
Chemical Name or Material Tetramethylammonium hexafluorophosphate
Show More Show Less

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H332-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 209-194-3

RTECSNumber : BS8000000

TSCA : No

Recommended Storage : Ambient temperatures