Alfa Aesar™ Tetramethylammonium hydroxide, 2.38% w/w aq. soln., Electronic Grade, 99.9999% (metals basis)

Manufacturer: Alfa Aesar™ 44940LU

Quantity 4 x 1 gal.

 View more versions of this product

Catalog No. AA44940LU

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75-59-2
    Molecular Formula C4H13NO
    Linear Formula (CH3)4NOH
    Formula Weight 91.15
    MDL Number MFCD00008280
    UN Number UN1835
    Beilstein 3558708
    Merck Index 14,9224
    Chemical Name or Material Tetramethylammonium hydroxide
    Grade Electronic Grade

    Specifications

    Formula Weight 91.15
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 75-59-2
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: TETRAMETHYLAMMONIUM HYDROXIDE SOLUTION
    MDL Number MFCD00008280
    Beilstein 3558708
    Health Hazard 1 Danger
    Quantity 4 x 1 gal.
    Merck Index 14,9224
    Sensitivity Air Sensitive
    Linear Formula (CH3)4NOH
    UN Number UN1835
    Color Colorless to pale yellow
    Concentration 2.38% w/w aq. soln.
    TSCA TSCA
    Physical Form Liquid
    Molecular Formula C4H13NO
    RTECS Number PA0875000
    Health Hazard 2 GHS H Statement
    H300-H310-H370-H372-H314
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes damage to organs.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Chemical Name or Material Tetramethylammonium hydroxide
    Assay 99.9999%, (metals basis)
    Grade Electronic Grade

    GHS H Statement
    H300-H310-H370-H372-H314
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes damage to organs.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit