Tetramethylammonium hydroxide, 25% w/w aq. soln., Electronic Grade, 99.9999% (metals basis)

Manufacturer: Alfa Aesar 20932LU

Quantity 4 x 1 gal

 View more versions of this product

Catalog No. AA20932LU

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75-59-2
    Molecular Formula C4H13NO
    Linear Formula (CH3)4NOH
    Formula Weight 91.15
    MDL Number MFCD00008280
    UN Number UN1835
    Beilstein 3558708
    Merck Index 14,9224
    Synonym N,N,N,-trimethylmethanaminium hydroxide
    Chemical Name or Material Tetramethylammonium hydroxide
    Grade Electronic Grade

    Specifications

    Formula Weight 91.15
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C4H13NO
    Synonym N,N,N,-trimethylmethanaminium hydroxide
    Physical Form Liquid
    Beilstein 3558708
    Density 1.016g/mL
    Quantity 4 x 1 gal
    Linear Formula (CH3)4NOH
    CAS 75-59-2
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TETRAMETHYLAMMONIUM HYDROXIDE SOLUTION
    UN Number UN1835
    Merck Index 14,9224
    Sensitivity Air Sensitive
    TSCA TSCA
    Refractive Index 1.3840
    MDL Number MFCD00008280
    Health Hazard 1 Danger
    Boiling Point ≈102°C
    Color Colorless to pale yellow
    Health Hazard 2 GHS H Statement
    H300-H310-H370-H372-H314
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes damage to organs.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    RTECS Number PA0875000
    Concentration 25% w/w aq. soln.
    Chemical Name or Material Tetramethylammonium hydroxide
    Assay 99.9999%, (metals basis)
    Grade Electronic Grade

    GHS H Statement
    H300-H310-H370-H372-H314
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes damage to organs.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit