Learn More
Tetramethylammonium hydroxide, 25% w/w aq. soln., Electronic Grade, 99.9999% (metals basis), Thermo Scientific Chemicals
Description
Tetramethylammonium hydroxide is used to produce tetramethyl-ammonium azide. It is used as an anisotropic etchant of silicon, basic solvent in the development of acidic photoresist in photolithography process, as surfactant in the synthesis of ferrofluid and as a polarographic reagent. It finds application in the production of organic silicon, where it is used for computer silicon wafer surface brightener and cleaning agent. It is also involved in the purification of some metallic elements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Density | 1.016 g/mL |
Boiling Point | ∼102°C |
Refractive Index | 1.384 |
Linear Formula | (CH3)4NOH |
Quantity | 1 L |
UN Number | 1835 |
Beilstein | 3558708 |
Sensitivity | Air Sensitive |
Merck Index | 14,9224 |
Show More |
Safety and Handling
Hazard Category | H301-H310-H314-H370-H372 |
Hazard Statement | GHS H Statement H300-H310-H370-H372-H314 Fatal if swallowed. Fatal in contact with skin. Causes damage to organs. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. |
Precautionary Statement | P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TETRAMETHYLAMMONIUM HYDROXIDE SOLUTION |
EINECSNumber | 200-882-9 |
RTECSNumber | PA0875000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.