Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium Hydroxide, 5-Hydrate, BAKER™, J.T.Baker™

Click to view available options
Quantity:
25g
1kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

10424-65-4
C4H23NO6
181.229
MYXKPFMQWULLOH-UHFFFAOYSA-M
82620

A grade of chemicals for laboratory use

Specific Gravity 1L = 1.00kg
Quantity 25g
Packaging Wide Mouth Amber Glass Bottle
SMILES C[N+](C)(C)C.O.O.O.O.O.[OH-]
IUPAC Name tetramethylazanium;hydroxide;pentahydrate
Formula Weight 181.23
Chemical Name or Material Tetramethylammonium Hydroxide, 5-Hydrate