Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium Hydroxide Pentahydrate 97.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
500g
10424-65-4
C4H23NO6
181.229
MFCD00149566
MYXKPFMQWULLOH-UHFFFAOYSA-M
tetramethylammonium hydroxide pentahydrate,b2hkt2lckq,unii-b2hkt2lckq,methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate,tmaoh,tetramethylammoniumhydroxid,tetrametanoamonio hydroxido,tetrametiloamonio hydroxido,methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5,acmc-2098bm
82620
tetramethylazanium;hydroxide;pentahydrate
C[N+](C)(C)C.O.O.O.O.O.[OH-]
10424-65-4
C4H23NO6
181.229
MFCD00149566
MYXKPFMQWULLOH-UHFFFAOYSA-M
tetramethylammonium hydroxide pentahydrate,b2hkt2lckq,unii-b2hkt2lckq,methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate,tmaoh,tetramethylammoniumhydroxid,tetrametanoamonio hydroxido,tetrametiloamonio hydroxido,methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5,acmc-2098bm
82620
tetramethylazanium;hydroxide;pentahydrate
C[N+](C)(C)C.O.O.O.O.O.[OH-]
Quantity 500g
Melting Point 73°C
Color White
UN Number 3423
Formula Weight 91.15
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material Tetramethylammonium Hydroxide Pentahydrate

EINECSNumber : (2)-0186

RTECSNumber : PA0875000

TSCA : Yes