Learn More
Tetramethylammonium perchlorate, Thermo Scientific Chemicals
Description
Tetramethylammonium perchlorate acts as an intermediate in organic and chemical synthesis. It is used as a mobile phase in high performance liquid chromatographic analysis and in the quantitation of olanzapine clozapine and N-desmethylclozapine. It acts as a supporting electrolyte in the electrochemical reduction of electrochemical reduction of 1-bromodecanr, 1-iododecane,2-iodooctane and 2-bromooctane at mercury cathodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C |
Quantity | 100 g |
UN Number | UN1479 |
Solubility Information | Soluble in water. |
Formula Weight | 173.6 |
Physical Form | Crystalline |
Chemical Name or Material | Tetramethylammonium perchlorate |
Safety and Handling
Hazard Category | H272-H315-H319-H335 |
Hazard Statement | GHS H Statement H272-H315-H319-H335 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S. |
EINECSNumber | 219-805-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.