Alfa Aesar™ Tetramethylcyclopentadiene, mixed isomers, tech. 85%

Manufacturer: Alfa Aesar™ 3857714

Quantity 25g

 View more versions of this product

Catalog No. AA3857714

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 4249-10-9
    Formula Weight 122.21
    MDL Number MFCD00145385
    UN Number UN3295
    Chemical Name or Material Tetramethylcyclopentadiene, Mixed isomers
    Grade Technical Grade

    Specifications

    Formula Weight 122.21
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Density 0.808g/mL
    CAS 4249-10-9
    UN Number UN3295
    Flash Point 42° (108°F)
    Quantity 25g
    Physical Form Liquid
    Health Hazard 1 Warning
    Boiling Point 55° to 59°C (13mmHg)
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    MDL Number MFCD00145385
    Refractive Index 1.4720
    Sensitivity Air & Moisture Sensitive
    Color Colorless
    Chemical Name or Material Tetramethylcyclopentadiene, Mixed isomers
    Assay 85%
    Grade Technical Grade

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit