Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylene sulfone, 99+%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
126-33-0
C4H8O2S
120.17
HXJUTPCZVOIRIF-UHFFFAOYSA-N
sulfolane,tetramethylene sulfone,tetrahydrothiophene 1,1-dioxide,sulfolan,sulpholane,sulfalone,sulphoxaline,1,1-dioxothiolan,dioxothiolan,thiophan sulfone
31347
CHEBI:74794
thiolane 1,1-dioxide
C1CCS(=O)(=O)C1
126-33-0
C4H8O2S
120.17
HXJUTPCZVOIRIF-UHFFFAOYSA-N
sulfolane,tetramethylene sulfone,tetrahydrothiophene 1,1-dioxide,sulfolan,sulpholane,sulfalone,sulphoxaline,1,1-dioxothiolan,dioxothiolan,thiophan sulfone
31347
CHEBI:74794
thiolane 1,1-dioxide
C1CCS(=O)(=O)C1
Viscosity 10.34 mPa.s (30°C)
Specific Gravity 1.26
Boiling Point 285.0°C
Melting Point 27.4°C to 27.8°C
Color Yellow
Water 0.3% max.
Density 1.2600g/mL
Quantity 1L
Sulfated Ash 0.1% max.
Refractive Index 1.4820 to 1.4850
Flash Point 165°C
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Beilstein 17,I,5
Fieser 01,1144; 02,402; 05,651; 13,256
Merck Index 15,9085
Solubility Information Solubility in water: soluble. Other solubilities: soluble in diluted mineral acids, soluble in acetone, benzene and toluene, freely soluble in alcohol
Formula Weight 120.17
Physical Form Liquid After Melting
Percent Purity 99+%
Chemical Name or Material Tetramethylene sulfone
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 204-783-1