Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylene sulfoxide, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1600-44-8
C4H8OS
104.17
MFCD00005477
ISXOBTBCNRIIQO-UHFFFAOYSA-N
tetramethylene sulfoxide,tetrahydrothiophene 1-oxide,thiophane oxide,tetrahydrothiophene oxide,thiophane monoxide,thiophane 1-oxide,thiophene, tetrahydro-, 1-oxide,tetramethylene sulphoxide,thiophene, tetrahydro, 1-oxide,tetrametylene sulfoxide
1128
thiolane 1-oxide
C1CCS(=O)C1
1600-44-8
C4H8OS
104.17
MFCD00005477
ISXOBTBCNRIIQO-UHFFFAOYSA-N
tetramethylene sulfoxide,tetrahydrothiophene 1-oxide,thiophane oxide,tetrahydrothiophene oxide,thiophane monoxide,thiophane 1-oxide,thiophene, tetrahydro-, 1-oxide,tetramethylene sulphoxide,thiophene, tetrahydro, 1-oxide,tetrametylene sulfoxide
1128
thiolane 1-oxide
C1CCS(=O)C1
Flash Point >112°C
Infrared Spectrum Authentic
Specific Gravity 1.15
Quantity 100g
Density 1.1500g/mL
Refractive Index 1.5180 to 1.5220
Packaging Glass bottle
Color Colorless to Yellow
Boiling Point 235.0°C to 237.0°C
Assay Percent Range 97%
Fieser 01,1145
Formula Weight 104.17
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Tetramethylene sulfoxide, 98+%
Show More Show Less

EINECSNumber : 216-493-2

RTECSNumber : XN0830000

TSCA : TSCA