Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals

Catalog Number p-198449
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
1600-44-8
C4H8OS
104.17
MFCD00005477
ISXOBTBCNRIIQO-UHFFFAOYSA-N
tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide
1128
thiolane 1-oxide
C1CCS(=O)C1
1600-44-8
C4H8OS
104.17
MFCD00005477
ISXOBTBCNRIIQO-UHFFFAOYSA-N
tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide
1128
thiolane 1-oxide
C1CCS(=O)C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 235.0°C to 237.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5180 to 1.5220
Quantity 5 g
Fieser 01,1145
Specific Gravity 1.15
Formula Weight 104.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tetramethylene sulfoxide, 98+%
Show More Show Less
EINECSNumber 216-493-2
RTECSNumber XN0830000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit