Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylpyrazine, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
1124-11-4
C8H12N2
136.198
MFCD00006146
FINHMKGKINIASC-UHFFFAOYSA-N
tetramethylpyrazine,ligustrazine,pyrazine, tetramethyl,bs factor,tetramethylpyrazin,liqustrazine,tetrapyrazine,2,3,5,6-tetramethyl pyrazine,chuanxiongzine,ligustizine
14296
2,3,5,6-tetramethylpyrazine
CC1=C(N=C(C(=N1)C)C)C
1124-11-4
C8H12N2
136.198
MFCD00006146
FINHMKGKINIASC-UHFFFAOYSA-N
tetramethylpyrazine,ligustrazine,pyrazine, tetramethyl,bs factor,tetramethylpyrazin,liqustrazine,tetrapyrazine,2,3,5,6-tetramethyl pyrazine,chuanxiongzine,ligustizine
14296
2,3,5,6-tetramethylpyrazine
CC1=C(N=C(C(=N1)C)C)C
Color White
Infrared Spectrum Authentic
Density 1.0800g/mL
Packaging Glass bottle
Quantity 25g
Melting Point 82°C to 86°C
Assay Percent Range 98%
Beilstein 23, 99
Formula Weight 136.2
Physical Form Fine Crystals
Percent Purity ≥97.5%
Chemical Name or Material Tetramethylpyrazine, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 214-391-2

RTECSNumber : UQ3905000

TSCA : TSCA