Learn More
Molecular Probes™ Tetramethylrhodamine-6-Isothiocyanate (6-TRITC; R isomer) Non-distribution product as customer accommodation.
tetramethylrhodamine-6-isothiocyanate (6-TRITC; R isomer)
Manufacturer:  Molecular Probes™ T1481
Catalog No. T1481
Add to cart
Description
Description
The amine-reactive tetramethylrhodamine-6-isothiocyanate (6-TRITC; R isomer) can be used to can be used to create bright red-orange fluorescent bioconjugates with excitation/emission maxima ~555/580 nm.
Specifications
Specifications
Tetramethylrhodamine-6-Isothiocyanate (6-TRITC; R isomer), 5mg, Purple, Solid, Amine Reactivity, 80724-20-5 CAS Number, C25H21N3O3S Molecular Formula, 443.51 Molecular Weight, 580nm Emission, ∽555nm Excitation, –5 to –30°C Storage Req
Purple
TMR (Tetramethylrhodamine), TRITC Isomers
–5 to –30°C
443.51kDa
580nm
Solid
Labeling chemistry, protein labeling and crosslinking, protein biology, cell analysis, labeling amines, amine-reactive quenchers, protein, peptide and antibody labeling, labeling proteins and antibodies with fluorophores
Tetramethylrhodamine-6-Isothiocyanate (6-TRITC; R isomer)
5mg
∽555nm
98% by HPLC, 99% by HPLC Isomer
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.