Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylsilane, NMR Grade, 99.9+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
75-76-3
C4H12Si
88.22
MFCD00008274
CZDYPVPMEAXLPK-UHFFFAOYSA-N
silane, tetramethyl,tetramethylsilicane,tetramethyl silane,silicon, tetramethyl,tetramethyl-silane,unii-41y0rbg14q,me4si,ch3 4si,si ch3 4,chembl68073
6396
CHEBI:85361
tetramethylsilane
C[Si](C)(C)C
75-76-3
C4H12Si
88.22
MFCD00008274
CZDYPVPMEAXLPK-UHFFFAOYSA-N
silane, tetramethyl,tetramethylsilicane,tetramethyl silane,silicon, tetramethyl,tetramethyl-silane,unii-41y0rbg14q,me4si,ch3 4si,si ch3 4,chembl68073
6396
CHEBI:85361
tetramethylsilane
C[Si](C)(C)C
Suitability (for 1H-NMR position reference) passes
Specific Gravity 0.64
Quantity 25g
Color Colorless
Density 0.6400g/mL
Flash Point −27°C
Boiling Point 26.0°C to 28.0°C
Melting Point -99.0°C
Refractive Index 1.3585
Packaging Glass bottle
Assay Percent Range 99.9+%
Linear Formula Si(CH3)4
Beilstein 04, 625
Merck Index 15, 9377
Solubility Information Solubility in water: 0.02g/L (25°C). Other solubilities: soluble in most common organic solvents
Formula Weight 88.22
Physical Form Volatile Liquid
Percent Purity ≥99.9%
Chemical Name or Material Tetramethylsilane, 99.9+%
Show More Show Less

GHS H Statement:
Extremely flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-899-1

TSCA : TSCA