Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethyltetraselenafulvalene, 97%, ACROS Organics™

Click to view available options
Quantity:
10mg
50mg
250mg
Packaging:
Glass bottle
55259-49-9
C10H12Se4
448.04
MFCD00011507
YMWLPMGFZYFLRP-UHFFFAOYSA-N
tetramethyltetraselenafulvalene,tmtsf,1,3-diselenole, 2-4,5-dimethyl-1,3-diselenol-2-ylidene-4,5-dimethyl,2-4,5-dimethyl-1,3-diselenol-2-ylidene-4,5-dimethyl-1,3-diselenole,tetramethyltetraselenafulvalene, purum,4,4',5,5'-tetramethyl-.delta.2,2'-bi-1,3-diselenole,1,3-diselenole, 4,5-dimethyl-2-4,5-dimethyl-1,3-diselenol-2-ylidene
143265
2-(4,5-dimethyl-1,3-diselenol-2-ylidene)-4,5-dimethyl-1,3-diselenole
CC1=C([Se]C(=C2[Se]C(=C([Se]2)C)C)[Se]1)C
55259-49-9
C10H12Se4
448.04
MFCD00011507
YMWLPMGFZYFLRP-UHFFFAOYSA-N
tetramethyltetraselenafulvalene,tmtsf,1,3-diselenole, 2-4,5-dimethyl-1,3-diselenol-2-ylidene-4,5-dimethyl,2-4,5-dimethyl-1,3-diselenol-2-ylidene-4,5-dimethyl-1,3-diselenole,tetramethyltetraselenafulvalene, purum,4,4',5,5'-tetramethyl-.delta.2,2'-bi-1,3-diselenole,1,3-diselenole, 4,5-dimethyl-2-4,5-dimethyl-1,3-diselenol-2-ylidene
143265
2-(4,5-dimethyl-1,3-diselenol-2-ylidene)-4,5-dimethyl-1,3-diselenole
CC1=C([Se]C(=C2[Se]C(=C([Se]2)C)C)[Se]1)C
Color Red-Violet
Melting Point 265.0°C to 270.0°C
Quantity 250mg
Packaging Glass bottle
NMR Complies, 96.0% min.
Assay Percent Range 97%
Formula Weight 448.04
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Tetramethyltetraselenafulvalene, 97%

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger