Alfa Aesar™ Tetramethyltin, 98%

Manufacturer: Alfa Aesar™ 7114504

Quantity 2g

 View more versions of this product

Catalog No. AA7114504

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 594-27-4
    Molecular Formula (CH3)4Sn
    Formula Weight 178.83
    MDL Number MFCD00008278
    UN Number UN3384
    Chemical Name or Material Tetramethyltin

    Specifications

    Formula Weight 178.83
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point −12°C (9°F)
    Health Hazard 2 GHS H Statement
    H225-H300-H310-H330
    Highly flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    TSCA TSCA
    MDL Number MFCD00008278
    CAS 594-27-4
    UN Number UN3384
    Density 1.291g/mL
    Refractive Index 1.4410
    Boiling Point 74° to 75°C
    Melting Point −54°C
    Physical Form Liquid
    Molecular Formula (CH3)4Sn
    Solubility Information Insoluble in water; Soluble in organic solvents
    Odor Unpleasant
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
    RTECS Number WH8630000
    Quantity 2g
    Health Hazard 1 Danger
    Chemical Name or Material Tetramethyltin
    Assay 98%

    GHS H Statement
    H225-H300-H310-H330
    Highly flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit