Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylarsonium chloride monohydrate, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
104170-16-3
C24H22AsClO
436.811
MFCD00149995
NGDWSDTZKCSLRK-UHFFFAOYSA-M
tetraphenylarsenic chloride,tetraphenylarsonium chloride monohydrate,arsonium, tetraphenyl-,chloride, hydrate 1:1:1,phenylarsonium chloride,tetraphenylarsanium chloride hydrate,acmc-20m6yd,c24h20as.cl.h2o,tetraphenyl-arsonium hydrate chloride,tetraphenylarsonium v chloride hydrate
2723786
tetraphenylarsanium;chloride;hydrate
C1=CC=C(C=C1)[As+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.O.[Cl-]
104170-16-3
C24H22AsClO
436.811
MFCD00149995
NGDWSDTZKCSLRK-UHFFFAOYSA-M
tetraphenylarsenic chloride,tetraphenylarsonium chloride monohydrate,arsonium, tetraphenyl-,chloride, hydrate 1:1:1,phenylarsonium chloride,tetraphenylarsanium chloride hydrate,acmc-20m6yd,c24h20as.cl.h2o,tetraphenyl-arsonium hydrate chloride,tetraphenylarsonium v chloride hydrate
2723786
tetraphenylarsanium;chloride;hydrate
C1=CC=C(C=C1)[As+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.O.[Cl-]
Phase transfer catalyst
Quantity 10g
Packaging Glass bottle
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula (C6H5)4AsCl·H2O
Formula Weight 436.82
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material Tetraphenylarsonium chloride monohydrate, 97%

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger