Alfa Aesar™ Tetraphenyllead, 97%

Manufacturer: Alfa Aesar™ A1220814

Quantity 25g

 View more versions of this product

Catalog No. AAA1220814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 595-89-1
    Molecular Formula (C6H5)4Pb
    Formula Weight 515.61
    MDL Number MFCD00002999
    UN Number UN3467
    Chemical Name or Material Tetraphenyllead

    Specifications

    Formula Weight 515.61
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula (C6H5)4Pb
    CAS 595-89-1
    Boiling Point 126°C (13mmHg)
    UN Number UN3467
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
    Melting Point 227° to 228°C
    Quantity 25g
    MDL Number MFCD00002999
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Tetraphenyllead
    Assay 97%

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit