Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylphosphonium Iodide 98.0+%, TCI America™

Click to view available options
Quantity:
10g
25g
2065-67-0
C24H20IP
466.302
MFCD00011917
AEFPPQGZJFTXDR-UHFFFAOYSA-M
tetraphenylphosphonium iodide,tetraphenylphosphoniumiodide,tetraphenylphosphanium iodide,acmc-1cpca
2724164
tetraphenylphosphanium;iodide
C1=CC=C(C=C1)[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[I-]
2065-67-0
C24H20IP
466.302
MFCD00011917
AEFPPQGZJFTXDR-UHFFFAOYSA-M
tetraphenylphosphonium iodide,tetraphenylphosphoniumiodide,tetraphenylphosphanium iodide,acmc-1cpca
2724164
tetraphenylphosphanium;iodide
C1=CC=C(C=C1)[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[I-]
Color White-Yellow
Quantity 10g
Formula Weight 466.30
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Tetraphenylphosphonium Iodide

EINECSNumber : (3)-2500

TSCA : No