Alfa Aesar™ Tetraphenylthiophene, 97%

Manufacturer: Alfa Aesar™ B2258803

Quantity 1g

 View more versions of this product

Catalog No. AAB2258803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1884-68-0
    Molecular Formula C28H20S
    Formula Weight 388.53
    MDL Number MFCD00014527
    Beilstein 297707
    Chemical Name or Material Tetraphenylthiophene

    Specifications

    Formula Weight 388.53
    Molecular Formula C28H20S
    MDL Number MFCD00014527
    CAS 1884-68-0
    Beilstein 297707
    Melting Point 178° to 186°C
    Quantity 1g
    Chemical Name or Material Tetraphenylthiophene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit