Alfa Aesar™ Tetraphenyltin

Manufacturer: Alfa Aesar™ 7111722

Quantity 100g

 View more versions of this product

Catalog No. AA7111722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 595-90-4
    Molecular Formula (C6H5)4Sn
    Formula Weight 427.11
    MDL Number MFCD00003006
    UN Number UN3146
    Synonym Tetraphenylstannane
    Chemical Name or Material Tetraphenyltin

    Specifications

    Formula Weight 427.11
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00003006
    CAS 595-90-4
    Quantity 100g
    Boiling Point >420°C
    TSCA TSCA
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Melting Point 224° to 227°C
    Molecular Formula (C6H5)4Sn
    Synonym Tetraphenylstannane
    Odor Characteristic
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
    Physical Form Crystalline
    UN Number UN3146
    Health Hazard 1 Danger
    Chemical Name or Material Tetraphenyltin

    GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit