Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrindole mesylate, Tocris Bioscience™

Catalog Number 072310
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
170964-68-8
C21H30N2O3S
390.542
CWPNPYVNPOAMHY-UHFFFAOYSA-N
tetrindole mesylate, tetrindole methanesulfonate, tetrindol, aronis25109, methanesulfonic acid; tetrindole, 2,3,3a,4,5,6-hexahydro-8-cyclohexyl-1h-pyrazino 3,2,1-j,k carbazole mesylate, 8-cyclohexyl-2,3,3a,4,5,6-hexahydro-1h-pyrazino 3,2,1-jk carbazole methanesulfonate, rac-8-cyclohexyl-2,3,3a,4,5,6-hexahydro-1h-pyrazino 3,2,1-jk carbazole methanesulfonate, rac-8-cyclohexyl-2,3,3a,4,5,6-hexahydro-1h-pyrazino 3,2,1-jk carbazol-3-ium methanesulfonate
54792409
CHEBI:77779
CS(=O)(=O)O.C1CCC(CC1)C2=CC3=C(C=C2)N4CCNC5C4=C3CCC5
This item is not returnable. View return policy
170964-68-8
C21H30N2O3S
390.542
CWPNPYVNPOAMHY-UHFFFAOYSA-N
tetrindole mesylate, tetrindole methanesulfonate, tetrindol, aronis25109, methanesulfonic acid; tetrindole, 2,3,3a,4,5,6-hexahydro-8-cyclohexyl-1h-pyrazino 3,2,1-j,k carbazole mesylate, 8-cyclohexyl-2,3,3a,4,5,6-hexahydro-1h-pyrazino 3,2,1-jk carbazole methanesulfonate, rac-8-cyclohexyl-2,3,3a,4,5,6-hexahydro-1h-pyrazino 3,2,1-jk carbazole methanesulfonate, rac-8-cyclohexyl-2,3,3a,4,5,6-hexahydro-1h-pyrazino 3,2,1-jk carbazol-3-ium methanesulfonate
54792409
CHEBI:77779
CS(=O)(=O)O.C1CCC(CC1)C2=CC3=C(C=C2)N4CCNC5C4=C3CCC5

MAO-A inhibitor

Novel antidepressant, a selective inhibitor of MAO-A.
Quantity 10 mg
Formula Weight 390.54
Percent Purity >99%
Chemical Name or Material Tetrindole mesylate
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit