Tetronic acid, 96%

Manufacturer: Alfa Aesar A1407906

Quantity 5g

 View more versions of this product

Catalog No. AAA1407906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4971-56-6
    Molecular Formula C4H4O3
    Formula Weight 100.07
    MDL Number MFCD00010086
    Beilstein 108442
    Chemical Name or Material Tetronic Acid

    Specifications

    Formula Weight 100.07
    Quantity 5g
    MDL Number MFCD00010086
    Beilstein 108442
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for se
    Assay Percent Range 96%
    CAS 4971-56-6
    Molecular Formula C4H4O3
    Chemical Name or Material Tetronic Acid
    Assay 96%

    Description

    Applications
    Tetronic acid is used as a precursor of a variety of substituted and ring-fused furans and butenolides. It is an acidic building block for heterocyclic compounds for medicinal applications. It is used for selective O-alkyaltion reactions involving cesium fluoride. It is also used to prepare 3H-furo[3,4-b]quinolin-1-one by reacting with 2-amino-benzaldehyde.

    Solubility
    Soluble in water, dimethyl sulfoxide, acetonitrile and alcohol.

    Notes
    Incompatible with strong oxidizing agents and strong alkalis (bases).

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for se

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit