Learn More
BioVision IncSupplier Diversity Partner TEV Protease Activity Assay Kit (Fluorometric): Simple kit with a fast protocol to fluorometrically (Ex/Em = 490/560 nm) detect as low as 50 ng/well of TEV Protease activity in viral lysates and purified protein samples. 100 assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K842100
TEV Protease (EC: 3.4.22.44, Tobacco Etch Virus protease) is a cysteine protease that recognizes the cleavage site of Glu-Xaa- Xaa-Y- Xaa-Gln-(Gly/Ser) and cleaves between Gln and Gly/Ser. The optimal sequence is Glu-Asn-Leu-Tyr-Phe-Gln-Ser/Glycine (ENLYFQS/G). TEV Protease has high specificity and great stability and is active over a wide range of temperatures (4-37°C) with an optimal activity at 34°C. BioVision’s TEV Protease Activity assay kit utilizes the ability of TEV Protease to cleave a synthetic Fluorescein-based peptide substrate to release fluorescein which can be easily quantified using a fluorometer or fluorescence microplate reader (Ex/Em = 490/560 nm). This assay kit is simple, rapid and can detect TEV Protease activity as low as 50 ng/well in viral lysates and purified protein samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9955