Learn More
Waldmann Lighting™ TEVISIO LED Magnifier Light, 3.5D + 8D, with Dimming and Segment Switching
Waldmann Lighting™ Tevisio and Televisiomax magnifier lights combine the latest in LED technology with magnification. Ideal for the lab, research, or any work surface where glare-free close-up work is necessary.
Manufacturer:  Waldmann Lighting™ 11291800000587332
 View more versions of this product
Catalog No. 11-000-115
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • 48 premium LEDs
  • 6" diameter wide-angle, scratch-resistant magnifying glass lens
  • Spring-balanced arm for easy adjustments
  • 3.5 diopter or 3.5d+8d magnification
  • Available in 31 in. or 39 in. arm length versions
  • 40% energy savings compared to fluorescents
  • 50,000 hours maintenance-free LED life
  • 100% to 10% dimming by touch
  • ETL/cETL approved
Specifications
Specifications
6 in.
15.24cm
Articulated Arm; 12-foot cord, plug; ETL/cETL, CE certified
39 in.
600 lumens
Silver
Anodized Aluminum
3.5 diopters
10.4 in. diameter x 0.8 in.
99.06 cm
3.5d+8d magnification