Learn More
TEX11 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 172661AP
Catalog No. 50-173-1479
Add to cart
Description
Description
This gene is X-linked and is expressed in only male germ cells. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
Specifications
TEX11
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IYF3
testis expressed 11, TEX11, TGC1, TSGA3
Rabbit
IgG
150 μL
-20°C
Primary
56159
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TEX11
Liquid
TEX11
TEX11 Fusion Protein Ag11012
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human