Learn More
TEX28 Monoclonal Antibody (1A8), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524399
Description
Peptide Sequence: MVLKAEHTRS PSATLPSNVP SCRSLSSSED GPSGPSSLAD GGLAHNLQDS VRHRILYLSE QLRVEKASRD GNTVSYLKLV SKADRHQVPH IQQAFEKVNQ RASATIAQIE HRLHQCHQQL QELEEGCRPE GLLLMAESDP ANCEPPSEKA LLSEPPEPGG EDGPVNLPHA SRPFILESRF QSLQQGTCLE TEDVAQQQNL LLQKVKAELE EAKRFHISLQ ESYHSLKERS LTDLQLLLES LQEEKCRQAL MEEQVNGRLQ GQLNEIYNLK HNLACSEERM AYLSYERAKE IWEITETFKS RISKLEMLQQ VTQLEAAEHL QSRPPQMLFK FLSPRLSLAT VLLVFVSTLC ACPSSLISSR LCTCTMLMLI GLGVLAWQRW RAIPATDWQE WVPSRCRLYS KDSGPPADGP
Specifications
TEX28 | |
Monoclonal | |
Unconjugated | |
TEX28 | |
CXorf2, TEX28P1, TEX28P2, fTEX, testis-specific protein TEX28 | |
Mouse | |
Protein A | |
RUO | |
1527 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1A8 | |
PBS with no preservative; pH 7.4 | |
O15482 | |
TEX28 | |
TEX28 (AAH26183, 1 a.a. ∽ 410 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.