Learn More
Texas Instruments™ TI-84 Plus and Plus Silver Calculators Products for Science Education
Preloaded with interactive geometry software and more.
$127.00
Specifications
Includes USB computer cable
Port Type I/O and USB
Power Supply 4 AAA with Lithium battery backup
Screen Size 64 x 96
Display Type Digital
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S68601 Products for Science Education
View Documents
Texas Instruments Inc
TI84PLUS
Graphing Calculator
Each for $127.00
Add to cart
Description
Description
Extensive graphing and functions like parameter, polar, sequence, choice of graph formats, logic operations, interactive equation solver, matrices and more.
Both models feature:
  • Data analysis includes scatter plots and statistics
  • Share data with the TI-83 Plus
  • Mini USB cable enables instant computer connectivity
  • Connect to ViewScreen™ panel (S66344) or TI-Presenter™ projection devices
  • Powered by four AAA batteries with lithium battery backup
  • Automatic Power Down™ (APD) capabilities
Plus Silver Model also features:
  • Additional preloaded application software for periodic table; probability simulations; inequality, conic and transform graphing; study cards; and science tools
  • Complete electronic guidebook
  • AP biology review questions on CD-ROM
Specifications
  • 64 x 96 pixel display with split screen
  • 24K available memory Flash ROM: 480K for Plus; 1.5MB for Plus Silver
  • Clock
  • I/O and USB ports
Warranty and Services
One year limited
Specifications
Specifications
USB computer cable
4 AAA with Lithium battery backup
Digital
Graphing Calculator
I/O and USB
64 x 96
Pre-loaded software applications- all to help you gain an academic edge from pre-algebra through calculus, as well as biology, chemistry and physics