Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texas Instruments™ TI-Nspire™ CX Handheld — Teacher's Bundle and Pack
SDPSCI_ED

The perfect solution for the classroom, lab and field.

Supplier:  Texas Instruments™ TINSPCXTKYELLOW

Catalog No. S02280ND


Add to cart

Includes:
  • Teacher Bundle: One handheld, single-license TI-Nspire and CAS Teacher Software, rechargeable battery, wall adapter, USB unit-to-unit cable, USB unit-to-computer cable
  • Teacher Pack: Ten handhelds with rechargeable batteries installed, 90-day trial of TI-Nspire and CAS Teacher Software, ten USB cables, five unit-to-unit cables, one manual, docking station and poster

Description

Description

Explore dynamic simulations and collect data with Vernier™ sensors. A virtual and real-world way to explore science.

  • Explore hundreds of free TI-Nspire biology, chemistry, physics and earth science lessons and simulations from middle to high school grades at www.sciencenspired.com
  • Instantly turn the TI-Nspire CX handheld into a data collection device by attaching a Vernier EasyLink™ USB interface and sensor
  • For advanced data collection, use the TI-Nspire Lab Cradle (sold separately); supports simultaneous use of up to five Vernier sensors
  • Use digital images or your own photos to connect science concepts with real-world scenarios
  • Touchpad navigation works more like a laptop computer
  • Approved for standardized tests — SAT™, PSAT/NMSQT™, ACT™, IB and AP exams
  • Screen size: 320 × 240 pixels (3.2 in. [8.1cm] diagonal)
  • Screen resolution: 125 dpi; 16-bit color
  • Memory: 100MB storage memory/64MB operating memory

Order Info

Handhelds powered by the TI-Nspire rechargeable battery (included) that lasts up to two weeks of normal use on a single charge. No alkaline batteries needed.

Specifications

Specifications

Teacher Pack
320 x 240
Touchpad navigation
125dpi
6000g
Ten handhelds with rechargeable batteries installed, 90 day trial of TI-Nspire and CAS Teacher software, ten USB cables, five unit to unit cables, one manual, docking station and poster
Backlit LCD
Middle to High School
100MB storage memory/64MB operating memory
13.4 lb.
Explore hundreds of free TI-Nspire biology, chemistry, physics and earth science lessons and simulations
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'