Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texas Instruments™ TI-Nspire™ CX Handheld — Teacher's Bundle and Pack
SCI_ED

The perfect solution for the classroom, lab and field.

$2106.00

Specifications

Display Type Backlit LCD
For Use With (Application) Explore hundreds of free TI-Nspire biology, chemistry, physics and earth science lessons and simulations
Grade Middle to High School
Keypad Touchpad navigation
Memory 100MB storage memory/64MB operating memory
View More Specs

Includes:
  • Teacher Bundle: One handheld, single-license TI-Nspire and CAS Teacher Software, rechargeable battery, wall adapter, USB unit-to-unit cable, USB unit-to-computer cable
  • Teacher Pack: Ten handhelds with rechargeable batteries installed, 90-day trial of TI-Nspire and CAS Teacher Software, ten USB cables, five unit-to-unit cables, one manual, docking station and poster

Products 1
Catalog Number Mfr. No. Product Type Includes Weight (English) Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Includes Weight (English) Weight (Metric) Price Quantity & Availability  
S02280ND
SCI_ED
View Documents Promotion Details
Texas Instruments™
TINSPCXTKYELLOW
Teacher Pack Ten handhelds with rechargeable batteries installed, 90 day trial of TI-Nspire and CAS Teacher software, ten USB cables, five unit to unit cables, one manual, docking station and poster 13.4 lb. 6000g
Each for $2,106.00
Add to cart
 
Description

Description

Explore dynamic simulations and collect data with Vernier™ sensors. A virtual and real-world way to explore science.

  • Explore hundreds of free TI-Nspire biology, chemistry, physics and earth science lessons and simulations from middle to high school grades at www.sciencenspired.com
  • Instantly turn the TI-Nspire CX handheld into a data collection device by attaching a Vernier EasyLink™ USB interface and sensor
  • For advanced data collection, use the TI-Nspire Lab Cradle (sold separately); supports simultaneous use of up to five Vernier sensors
  • Use digital images or your own photos to connect science concepts with real-world scenarios
  • Touchpad navigation works more like a laptop computer
  • Approved for standardized tests — SAT™, PSAT/NMSQT™, ACT™, IB and AP exams
  • Screen size: 320 × 240 pixels (3.2 in. [8.1cm] diagonal)
  • Screen resolution: 125 dpi; 16-bit color
  • Memory: 100MB storage memory/64MB operating memory

Order Info

Handhelds powered by the TI-Nspire rechargeable battery (included) that lasts up to two weeks of normal use on a single charge. No alkaline batteries needed.

Specifications

Specifications

Backlit LCD
Middle to High School
100MB storage memory/64MB operating memory
320 x 240
Explore hundreds of free TI-Nspire biology, chemistry, physics and earth science lessons and simulations
Touchpad navigation
125dpi
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit