Learn More
Aat BioquestSupplier Diversity Partner Texas Red® hydrazide *Single Isomer* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 481
Texas Red® hydrazide reacts with carbonyl compounds such as aldehyde, ketones and acids to give bright red fluorescent conjugates. It is widely used for labeling glycoproteins and carbohydrates through reductive amination.
Catalog No. 50-195-2899